CAS RN 2963-77-1
CAS RN 2963-77-1
IUPAC Name:
4-(1H-1,3-benzodiazol-2-yl)aniline
Synonyms:
4-(1H-1,3-benzodiazol-2-yl)aniline
Molecular Weight (g/mol):
209.25
Molecular Formula:
C13H11N3
InChi Key:
VQFBXSRZSUJGOF-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1
1
–
1
of
1
results
1
4-(1H-Benzimidazol-2-yl)aniline 98.0+%, TCI America™
CAS: 2963-77-1 Molecular Formula: C13H11N3 Molecular Weight (g/mol): 209.252 MDL Number: MFCD00454395 InChI Key: VQFBXSRZSUJGOF-UHFFFAOYSA-N Synonym: 2-(4-Aminophenyl)benzimidazole PubChem CID: 345677 IUPAC Name: 4-(1H-benzimidazol-2-yl)aniline SMILES: C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N