Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-Benzimidazol-2-yl)aniline 98.0+%, TCI America™
SDP

Catalog No. B57105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B57105G 5 g
B57101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B57105G Supplier TCI America Supplier No. B57105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2963-77-1
Molecular Formula C13H11N3
Molecular Weight (g/mol) 209.252
MDL Number MFCD00454395
InChI Key VQFBXSRZSUJGOF-UHFFFAOYSA-N
Synonym 2-(4-Aminophenyl)benzimidazole
PubChem CID 345677
IUPAC Name 4-(1H-benzimidazol-2-yl)aniline
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N

Specifications

Melting Point 250°C
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1H-Benzimidazol-2-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.