accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOCH3OH3CCH3NO

CAS RN 29639-21-2

IUPAC Name: [2-(hexan-2-yl)-2-(11-methoxy-11-oxoundecyl)-4,4-dimethyl-1,3-oxazolidin-3-yl]oxidanyl
Synonyms: 2-(hexan-2-yl)-2-(11-methoxy-11-oxoundecyl)-4,4-dimethyl-1,3-oxazolidin-3-yloxidanyl
Molecular Weight (g/mol): 398.61
Molecular Formula: C23H44NO4
InChi Key: NSXKMYQRBHXAHX-UHFFFAOYNA-N
SMILES: CCCCC(C)C1(CCCCCCCCCCC(=O)OC)OCC(C)(C)N1[O]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results