Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-12-DOXYL-stearate, Free Radical, Thermo Scientific™

Catalog No. AC225110250
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225110250 25 mg
AC225110100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225110250 Supplier Thermo Scientific Chemicals Supplier No. 225110250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29639-21-2
Molecular Formula C23H44NO4
Molecular Weight (g/mol) 398.608
InChI Key JJBQJGSYLSZDNS-UHFFFAOYSA-N
Synonym methyl 12-doxyl-stearate, 12-doxylstearic acid methyl, methyl 11-2-hexyl-3, methyl 12-doxyl-stearate free radical, methyl 12-doxyl-stearate, free radical, 2-hexyl-2-11-methoxy-11-oxoundecyl-4,4-dimethyl-3-oxazolidinyloxy, 2-hexyl-2-10-methoxycarbonyl decyl-4,4-dimethyl-3-oxazolidinyloxy, 1-tert-butyloxycarbonyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-indazole, 2-hexyl-2-11-methoxy-11-oxoundecyl-4,4-dimethyl-3-oxazolidinyloxy, free radical
PubChem CID 2724320
IUPAC Name methyl 11-(2-hexyl-3-$l^{1}-oxidanyl-4,4-dimethyl-1,3-oxazolidin-2-yl)undecanoate
SMILES CCCCCCC1(N(C(CO1)(C)C)[O])CCCCCCCCCCC(=O)OC

Specifications

Quantity 25 mg
Formula Weight 384.57
Chemical Name or Material Methyl-12-DOXYL-stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.