accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOCH3OH

CAS RN 29681-21-8

IUPAC Name: 1-{2-[2-(propan-2-yloxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 1-[2-(2-isopropoxyethoxy)ethoxy]ethanol
Molecular Weight (g/mol): 192.26
Molecular Formula: C9H20O4
InChi Key: PMMZQEGTYWCTBY-UHFFFAOYNA-N
SMILES: CC(C)OCCOCCOC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results