Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Monoisopropyl Ether 98.0+%, TCI America™
SDP

Catalog No. T13735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13735G Supplier TCI America Supplier No. T13735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29681-21-8
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.26
MDL Number MFCD00145451
InChI Key PMMZQEGTYWCTBY-UHFFFAOYNA-N
Synonym Isopropyl Triglycol
PubChem CID 542224
IUPAC Name 1-{2-[2-(propan-2-yloxy)ethoxy]ethoxy}ethan-1-ol
SMILES CC(C)OCCOCCOC(C)O

Specifications

Color Colorless
Boiling Point 125°C
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monoisopropyl Ether
EINECSNumber (2)-0436&(7)-0097
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.