accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 29682-12-0

IUPAC Name: 1-[4-(benzyloxy)-2-hydroxyphenyl]ethan-1-one
Synonyms: 1-[4-(benzyloxy)-2-hydroxyphenyl]ethanone
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: AGQNLHOTLJFJCG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results