Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Benzyloxy-2'-hydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. B39501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39501G Supplier TCI America Supplier No. B39501G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29682-12-0
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00054612
InChI Key AGQNLHOTLJFJCG-UHFFFAOYSA-N
Synonym 1-4-benzyloxy-2-hydroxyphenyl ethanone, 4'-benzyloxy-2'-hydroxyacetophenone, 4-benzyloxy-2-hydroxyacetophenone, 2-hydroxy-4-benzyloxyacetophenone, ethanone, 1-2-hydroxy-4-phenylmethoxy phenyl, 1-2-hydroxy-4-phenylmethoxyphenyl ethanone, 1-2-hydroxy-4-phenylmethoxy-phenyl ethanone, 1-4-benzyloxy-2-hydroxyphenyl ethan-1-one
PubChem CID 309257
IUPAC Name 1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OCC2=CC=CC=C2)O

Specifications

Melting Point 105°C
Quantity 1 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Benzyloxy-2′-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.