accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClI

CAS RN 29682-41-5

IUPAC Name: 1,4-dichloro-2-iodobenzene
Synonyms: 1,4-dichloro-2-iodo-benzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: SBHVNORGKIPGCL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results