Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dichloro-2-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. D143525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D143525G Supplier TCI America Supplier No. D143525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29682-41-5
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00001036
InChI Key SBHVNORGKIPGCL-UHFFFAOYSA-N
Synonym 2,5-dichloroiodobenzene, benzene, 1,4-dichloro-2-iodo, 1,4-dichloro-2-iodo-benzene, pubchem3694, 2,5-dichlor-jodbenzol, 2,5-dichloro-iodobenzene, acmc-1ckh5, 2,5 dichloro 1-iodobenzol, 2-iodo-1,4-dichlorobenzene, ksc494k7h
PubChem CID 122452
IUPAC Name 1,4-dichloro-2-iodobenzene
SMILES ClC1=CC=C(Cl)C(I)=C1

Specifications

Quantity 25 g
Formula Weight 272.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dichloro-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.