accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2975-41-9

IUPAC Name: hydrogen 2,3-dihydro-1H-inden-2-amine chloride
Synonyms: 2-aminoindane
Molecular Weight (g/mol): 169.65
Molecular Formula: C9H12ClN
InChi Key: XEHNLVMHWYPNEQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1CC2=CC=CC=C2C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results