Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoindan 99.0+%, TCI America™
SDP

Catalog No. A13205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13205G 5 g
A132025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13205G Supplier TCI America Supplier No. A13205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2975-41-9
Molecular Formula C9H12ClN
Molecular Weight (g/mol) 169.65
MDL Number MFCD00082597
InChI Key XEHNLVMHWYPNEQ-UHFFFAOYSA-N
Synonym 2-aminoindan, 2-aminoindane, indan-2-amine, 2-indanamine, 2,3-dihydro-1h-inden-2-ylamine, indan-2-ylamine, indane-2-ylamine, 2-indanylamine, unii-1p810sq3ey, 1h-inden-2-amine, 2,3-dihydro
PubChem CID 76310
IUPAC Name hydrogen 2,3-dihydro-1H-inden-2-amine chloride
SMILES [H+].[Cl-].NC1CC2=CC=CC=C2C1

Specifications

Color Brown
Boiling Point 229°C
Quantity 5 g
UN Number 2735
Formula Weight 133.19
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminoindan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.