CAS RN 29767-20-2
IUPAC Name:
16-{[7,8-dihydroxy-2-(thiophen-2-yl)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Synonyms:
teniposide
Molecular Weight (g/mol):
656.66
Molecular Formula:
C32H32O13S
InChi Key:
NRUKOCRGYNPUPR-UHFFFAOYNA-N
SMILES:
COC1=CC(=CC(OC)=C1O)C1C2C(COC2=O)C(OC2OC3COC(OC3C(O)C2O)C2=CC=CS2)C2=CC3=C(OCO3)C=C12
1
–
3
of
3
results
1
R&D Systems™ Teniposide
A Tocris product, part of R&D Systems. DNA topoisomerase II inhibitor; viral transduction enhancer
2
Promotions Available
Thermo Scientific Chemicals Teniposide
Thermo Scientific Chemicals Teniposide
CAS: 29767-20-2 Molecular Formula: C32H32O13S
3
Teniposide 98.0+%, TCI America™
CAS: 29767-20-2 Molecular Formula: C32H32O13S