accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2978-11-2

IUPAC Name: N,N'-bis(propan-2-yl)[(4-nitrophenyl)methoxy]methanimidamide
Synonyms: N,N'-diisopropyl(4-nitrophenyl)methoxymethanimidamide
Molecular Weight (g/mol): 279.34
Molecular Formula: C14H21N3O3
InChi Key: FYDMCLCYYCIDMZ-UHFFFAOYSA-N
SMILES: CC(C)NC(OCC1=CC=C(C=C1)[N+]([O-])=O)=NC(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results