Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropyl-O-(4-nitrobenzyl)isourea 95.0+%, TCI America™
SDP

Catalog No. A55061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55061G Supplier TCI America Supplier No. A55061G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2978-11-2
Molecular Formula C14H21N3O3
Molecular Weight (g/mol) 279.34
MDL Number MFCD00042046
InChI Key FYDMCLCYYCIDMZ-UHFFFAOYSA-N
Synonym O-(4-Nitrobenzyl)-N,N′C-diisopropylisourea, 1,3-Diisopropyl-2-(4-nitrobenzyl)isourea
PubChem CID 76317
IUPAC Name (4-nitrophenyl)methyl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OCC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 44°C
Quantity 1 g
Formula Weight 279.34
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diisopropyl-O-(4-nitrobenzyl)isourea [for HPLC Labeling]
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.