CAS RN 298-57-7
IUPAC Name:
1-(diphenylmethyl)-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine
Synonyms:
cinnarizine aplexal cinnarizine, (e)-isomer cinnarizine l tartrate cinnarizine L-tartrate (1:1) cinazière stutgeron denapol cisaken dimitronal
Molecular Weight (g/mol):
368.52
Molecular Formula:
C26H28N2
InChi Key:
DERZBLKQOCDDDZ-JLHYYAGUSA-N
SMILES:
C(\C=C\C1=CC=CC=C1)N1CCN(CC1)C(C1=CC=CC=C1)C1=CC=CC=C1