Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnarizine, MP Biomedicals™

Catalog No. ICN15498625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15498625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15498625 Supplier MP Biomedicals, Inc Supplier No. 0215498625

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 298-57-7
Molecular Formula C26H28N2
Molecular Weight (g/mol) 368.524
InChI Key DERZBLKQOCDDDZ-JLHYYAGUSA-N
Synonym cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
PubChem CID 1547484
ChEBI CHEBI:31403
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Color White
Quantity 25 g
Formula Weight 368.52
Physical Form Powder
Chemical Name or Material Cinnarizine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.