accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOOONH2

CAS RN 29816-01-1

IUPAC Name: 2-(2-amino-N-methylacetamido)acetic acid
Synonyms: (2-amino-N-methylacetamido)acetic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: VYAMLSCELQQRAE-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)C(=O)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results