CAS RN 29875-06-7
IUPAC Name:
magnesium(2+) (3-methylphenyl)methanide chloride
Synonyms:
magnesium(2+) (3-methylphenyl)methanide chloride
Molecular Weight (g/mol):
164.92
Molecular Formula:
C8H9ClMg
InChi Key:
MPIXEEVXDGICED-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].CC1=CC=CC([CH2-])=C1
1
–
1
of
1
results
1
Sigma Aldrich 3,5-Dichlorophenylmagnesium Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.