accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29885-95-8

IUPAC Name: N-methylanilinium trifluoroacetate
Synonyms: N-methylanilinium trifluoroacetate
Molecular Weight (g/mol): 221.18
Molecular Formula: C9H10F3NO2
InChi Key: TXXJGCVOHDSYBC-UHFFFAOYSA-N
SMILES: [O-]C(=O)C(F)(F)F.C[NH2+]C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)