Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylanilinium Trifluoroacetate 99.0+%, TCI America™
SDP

Catalog No. M080125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M080125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M080125G Supplier TCI America Supplier No. M080125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29885-95-8
Molecular Formula C9H10F3NO2
MDL Number MFCD00043278
Synonym N-MATFA, N-Methylaniline Trifluoroacetate

Specifications

Melting Point 68°C
Color White
Quantity 25 g
Formula Weight 221.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylanilinium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.