accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OClClCl

CAS RN 299-84-3

IUPAC Name: O,O-dimethyl O-2,4,5-trichlorophenyl phosphorothioate
Synonyms: ronnel 芬氯磷 dermaphos caswell no. 724 C8H8cl3O3PS nanchor etrolene fenchlorphos korlan trichlormetaphos
Molecular Weight (g/mol): 321.53
Molecular Formula: C8H8Cl3O3PS
InChi Key: JHJOOSLFWRRSGU-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC(Cl)=C(Cl)C=C1Cl
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results