accessibility menu, dialog, popup

UserName

Viewing profile as user:
INO

CAS RN 299159-27-6

IUPAC Name: 4-[(4-iodophenyl)methyl]morpholine
Synonyms: 4-[(4-iodophenyl)methyl]morpholine
Molecular Weight (g/mol): 303.14
Molecular Formula: C11H14INO
InChi Key: IYDFKIKPNVVXQU-UHFFFAOYSA-N
SMILES: IC1=CC=C(CN2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results