accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2993-85-3

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl prop-2-enoate
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl prop-2-enoate
Molecular Weight (g/mol): 386.14
Molecular Formula: C10H6F12O2
InChi Key: QJEJDNMGOWJONG-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results