Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl acrylate

Catalog No. 4744285ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-442-85ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-442-85ML Supplier Sigma Aldrich Supplier No. 4744285ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl acrylate (DFHA) is a fluorous functional monomer that is majorly used in the surface functionalization. It can also be utilized in chemical vapor deposition process for the synthesis of nanomaterials.DFHA can be used in the preparation of hybrid fluorous monolithic columns for nano-liquid chromatography. It can be used in the surface modification of multi-walled carbon nanotubes (MWCNTs) for the formation of highly conductive composites.

Specifications

CAS 2993-85-3
Density 1.581 g/mL (at 25°C)
Boiling Point 197°C
Molecular Formula C10H6F12O2
Refractive Index n20/D 1.342 (literature)
Linear Formula H2C=CHCO2CH2(CF2)5 CHF2
Quantity 5 mL
MDL Number MFCD00080746
Synonym DFHA
Molecular Weight (g/mol) 386.13
Percent Purity 95%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.