accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2998-04-1

H2COOOOCH2

CAS RN 2998-04-1

IUPAC Name: 1,6-bis(prop-2-en-1-yl) hexanedioate
Synonyms: 1,6-bis(prop-2-en-1-yl) hexanedioate
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: FPODCVUTIPDRTE-UHFFFAOYSA-N
SMILES: C=CCOC(=O)CCCCC(=O)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results