Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl Adipate, TCI America™
SDP

Catalog No. A082425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A082425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A082425ML Supplier TCI America Supplier No. A082425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2998-04-1
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00039826
InChI Key FPODCVUTIPDRTE-UHFFFAOYSA-N
Synonym Adipic Acid Diallyl Ester
PubChem CID 18139
IUPAC Name bis(prop-2-enyl) hexanedioate
SMILES C=CCOC(=O)CCCCC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 149°C
Quantity 25 mL
Formula Weight 226.27
Physical Form Liquid
Chemical Name or Material Diallyl Adipate
RTECSNumber AV0350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.