accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONC

CAS RN 2999-46-4

IUPAC Name: ethyl 2-isocyanoacetate
Synonyms: ethyl 2-isocyanoacetate
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: FPULFENIJDPZBX-UHFFFAOYSA-N
SMILES: CCOC(=O)C[N+]#[C-]
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (8)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (4)
  • (5)