Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isocyanoacetate, 97%

Catalog No. AC299551000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299551000 100 g Glass Bottle
AC299550050 5 g Glass Bottle
AC299550250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC299551000 Supplier Thermo Scientific Chemicals Supplier No. 299551000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2999-46-4 | C5H7NO2 | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2999-46-4
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
InChI Key FPULFENIJDPZBX-UHFFFAOYSA-N
Synonym ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
PubChem CID 533707
IUPAC Name ethyl 2-isocyanoacetate
SMILES CCOC(=O)C[N+]#[C-]

Specifications

Color Amber to Brown
Density 1.0300g/mL
Boiling Point 194.0°C to 196.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4170 to 1.4250
Linear Formula CNCH2CO2C2H5
Quantity 100 g
Specific Gravity 1.03
Solubility Information Solubility in water: hydrolyses
Formula Weight 113.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl isocyanoacetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 221-077-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.