accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)ONH2OHHOII

CAS RN 300-39-0

IUPAC Name: (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Synonyms: 3,5-diiodotyrosine
Molecular Weight (g/mol): 432.98
Molecular Formula: C9H9I2NO3
InChi Key: NYPYHUZRZVSYKL-JLDDOWRYNA-N
SMILES: N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)