Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-L-tyrosine dihydrate, 99%, Thermo Scientific™

Catalog No. AC225611000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC225611000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225611000 Supplier Thermo Scientific Chemicals Supplier No. 225611000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 300-39-0
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C9H9I2NO3
Linear Formula HOC6H2I2CH2CH(NH2)COOH·2H2O
Quantity 100g
MDL Number MFCD00150275
Beilstein 14, IV, 2370
Specific Rotation Condition −20.70° (20°C c=2.5,0.1N NaOH/H2O)
Specific Rotation On dry substance: -17° to -20.7° (20°C, 589nm) (c=2.5, 0.1N NaOH/H2O)
Synonym 3,5-diiodo-l-tyrosine, l-3,5-diiodotyrosine, diiodotyrosine, l-diiodotyrosine, 3,5-diiodotyrosine, 3,5-l-diiodotyrosine, 3,5-iodo-l-tyrosine, h-tyr 3,5-i2-oh, dit amino acid, l-tyrosine, 3,5-diiodo
InChI Key NYPYHUZRZVSYKL-ZETCQYMHSA-N
SMILES C1=C(C=C(C(=C1I)O)I)CC(C(=O)O)N
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Molecular Weight (g/mol) 432.984
PubChem CID 9305
ChEBI CHEBI:15768
Formula Weight 469.01
Percent Purity ≥98.5% (on dry substance)
Chemical Name or Material 3, 5-Diiodo-L-tyrosine dihydrate, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-092-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.