CAS RN 300353-13-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Glucokinase Activator III, Ro-28-1675
A cell-permeable activator of glucokinase. Exhibits in vivo glucose-lowering and insulin-releasing activity