Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glucokinase Activator III, Ro-28-1675

Catalog No. 5096650001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-966-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-966-50001 Supplier MilliporeSigma™ Supplier No. 5096650001
Add to Cart
Add to Cart

A cell-permeable activator of glucokinase. Exhibits in vivo glucose-lowering and insulin-releasing activity

A cell-permeable thiazolyl-propanamide compound that acts as a glucokinase (GK) activator by increasing Vmax and decreasing [S]1/2 of GK-catalyzed reaction as well as freeing GK from the inhibitory action of GK regulatory protein (GKRP), resulting in increased. Induces GK nuclear-to-cytosol translocation in primary rat hepatocytes and reduces glucose concentration for insulin secretion stimulation from isolated rat pancreatic islets (threshold [glucose] = 3mm vs 6mm with or without 3μM Ro-28-1675). Exhibits in vivo glucose-lowering and insulin-releasing activity in non-diabetic C57BL/6 mice and Wistar rats and is efficacious in several murine and rodent type II diabetes mellitus (T2DM) models (10 to 50mg/kg via p.o.). Does not affect hexokinase I or II activity.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Glucokinase
CAS 300353-13-3
Molecular Formula C18H22N2O3S2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.