accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOOOCH3CH3H3C

CAS RN 3006-86-8

IUPAC Name: 1,1-bis(tert-butylperoxy)cyclohexane
Synonyms: 1,1-bis(tert-butylperoxy)cyclohexane
Molecular Weight (g/mol): 260.37
Molecular Formula: C14H28O4
InChi Key: HSLFISVKRDQEBY-UHFFFAOYSA-N
SMILES: CC(C)(C)OOC1(CCCCC1)OOC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)