Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Luperox™ 331M80, 1,1-Bis(tert-butylperoxy)cyclohexane solution

Catalog No. 531758500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-175-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-175-8500ML Supplier Sigma Aldrich Supplier No. 531758500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Luperox™ 331M80, 1,1-Bis(tert-butylperoxy)cyclohexane solution

Specifications

CAS 3006-86-8
Melting Point 65°C (SADT)
Density 0.891 g/mL (at 25°C)
Molecular Formula C14H28O4
Refractive Index n20/D 1.435
Linear Formula C6H10[OOC(CH3)3]2
Quantity 500 mL
MDL Number MFCD00084800
Synonym 1,1-Bis(tert-butylperoxy)cyclohexane solution
Molecular Weight (g/mol) 260.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.