CAS RN 300801-52-9
IUPAC Name:
1-(3-ethyl-5-methoxy-2,3-dihydro-1,3-benzothiazol-2-ylidene)propan-2-one
Synonyms:
1-(3-ethyl-5-methoxy-1,3-benzothiazol-2-ylidene)propan-2-one
Molecular Weight (g/mol):
249.33
Molecular Formula:
C13H15NO2S
InChi Key:
BGVLELSCIHASRV-UHFFFAOYSA-N
SMILES:
CCN1C(SC2=C1C=C(OC)C=C2)=CC(C)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Cdc2-Like Kinase Inhibitor, TG003
Thermo Scientific Chemicals Cdc2-Like Kinase Inhibitor, TG003
CAS: 300801-52-9 Molecular Formula: C13H15NO2S Synonym: Clk Inhibitor; TG003