accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 300829-22-5

IUPAC Name: 2-chloro-N-cyclopentyl-4-nitrobenzamide
Synonyms: 2-chloro-N-cyclopentyl-4-nitrobenzamide
Molecular Weight (g/mol): 268.7
Molecular Formula: C12H13ClN2O3
InChi Key: SWXOJSLQXXNFQE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)C(=O)NC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results