accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3009-97-0

IUPAC Name: 2-(phenylamino)acetonitrile
Synonyms: acetonitrile, anilino- (8CI)
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: KAXCEFLQAYFJKV-UHFFFAOYSA-N
SMILES: N#CCNC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)