Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylglycinonitrile, 97%, Thermo Scientific™

Catalog No. AC184500100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184500100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC184500100 Supplier Thermo Scientific Chemicals Supplier No. 184500100
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3009-97-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.16
MDL Number MFCD00044430
InChI Key KAXCEFLQAYFJKV-UHFFFAOYSA-N
Synonym 2-phenylamino acetonitrile, n-phenylglycinonitrile, anilinoacetonitrile, n-phenylglycinenitrile, phenylamino acetonitrile, glycinonitrile, n-phenyl, phenylglycine nitrile, 1-anilinoacetonitrile, n-cyanomethyl aniline, acetonitrile, anilino
PubChem CID 76372
IUPAC Name 2-anilinoacetonitrile
SMILES C1=CC=C(C=C1)NCC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHCH2CN
Quantity 10 g
Formula Weight 132.16
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N-Phenylglycinonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 221-129-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.