accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3N

CAS RN 3010-02-4

IUPAC Name: 2-(diethylamino)acetonitrile
Synonyms: acetonitrile, (diethylamino)-
Molecular Weight (g/mol): 112.18
Molecular Formula: C6H12N2
InChi Key: LVPZSMIBSMMLPI-UHFFFAOYSA-N
SMILES: CCN(CC)CC#N
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results