Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylaminoacetonitrile 98.0+%, TCI America™
SDP

Catalog No. D110425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D110425ML Supplier TCI America Supplier No. D110425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3010-02-4
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.18
MDL Number MFCD00001891
InChI Key LVPZSMIBSMMLPI-UHFFFAOYSA-N
Synonym diethylaminoacetonitrile, 2-diethylamino acetonitrile, acetonitrile, diethylamino, n,n-diethylaminoacetonitrile, nitril kiseliny diethylaminooctove, diethylamino acetonitrile, glycinonitrile, n,n-diethyl, unii-468yw0h2fa, acetonitrile, 2-diethylamino, nitril kiseliny diethylaminooctove czech
PubChem CID 44983
IUPAC Name 2-(diethylamino)acetonitrile
SMILES CCN(CC)CC#N

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 1992
Formula Weight 112.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylaminoacetonitrile
RTECSNumber AL8575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.