CAS RN 3010-57-9
IUPAC Name:
N,N-dimethyl-4-[2-(4-methylphenyl)diazen-1-yl]aniline
Synonyms:
N,N-dimethyl-4-[2-(4-methylphenyl)diazen-1-yl]aniline
Molecular Weight (g/mol):
239.32
Molecular Formula:
C15H17N3
InChi Key:
IPIZWPBCCYXHDW-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C)C=C1
1
–
1
of
1
results
1
4-(Dimethylamino)-4'-methylazobenzene 98.0+%, TCI America™
CAS: 3010-57-9 Molecular Formula: C15H17N3 Molecular Weight (g/mol): 239.322 MDL Number: MFCD00048015 InChI Key: IPIZWPBCCYXHDW-UHFFFAOYSA-N Synonym: 4-dimethylamino-4'-methylazobenzene,4'-methyl-p-dimethylaminoazobenzene,4'-methyl-4-dimethylaminoazobenzene,n,n-dimethyl-p-p-tolylazo aniline,n,n-dimethyl-4-p-tolylazo aniline,unii-610e3rbw8u,4'-methyl-4-dimethylamino azobenzene,p'-methyl-p-dimethylaminoazobenzene,aniline, n,n-dimethyl-4-p-tolylazo PubChem CID: 18168 IUPAC Name: N,N-dimethyl-4-[(4-methylphenyl)diazenyl]aniline SMILES: CC1=CC=C(C=C1)N=NC2=CC=C(C=C2)N(C)C