Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)-4'-methylazobenzene 98.0+%, TCI America™
SDP

Catalog No. M02085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02085G Supplier TCI America Supplier No. M02085G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3010-57-9
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.322
MDL Number MFCD00048015
InChI Key IPIZWPBCCYXHDW-UHFFFAOYSA-N
Synonym 4-dimethylamino-4'-methylazobenzene, 4'-methyl-p-dimethylaminoazobenzene, 4'-methyl-4-dimethylaminoazobenzene, n,n-dimethyl-p-p-tolylazo aniline, n,n-dimethyl-4-p-tolylazo aniline, unii-610e3rbw8u, 4'-methyl-4-dimethylamino azobenzene, p'-methyl-p-dimethylaminoazobenzene, aniline, n,n-dimethyl-4-p-tolylazo
PubChem CID 18168
IUPAC Name N,N-dimethyl-4-[(4-methylphenyl)diazenyl]aniline
SMILES CC1=CC=C(C=C1)N=NC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 239.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)-4′-methylazobenzene
RTECSNumber BX8400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.