accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CNHNOHOClSOO

CAS RN 30123-17-2

IUPAC Name: sodium 2-[(6-carboxyhexyl)amino]-6-chloro-10-methyl-9λ⁴-thia-10-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-9,9-bis(olate)
Synonyms: tianeptine
Molecular Weight (g/mol): 459.94
Molecular Formula: C21H25ClN2NaO4S
InChi Key: KBNZQVDKURREAX-UHFFFAOYNA-L
SMILES: [Na+].CN1C2=CC=CC=C2C(NCCCCCCC(O)=O)C2=CC=C(Cl)C=C2S1([O-])[O-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results