accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30131-16-9

IUPAC Name: 4-(4-phenylbutoxy)benzoic acid
Synonyms: 4-(4-phenylbutoxy)benzoic acid
Molecular Weight (g/mol): 270.33
Molecular Formula: C17H18O3
InChi Key: XWCWFMQMZZPALR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results