Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Phenylbutoxy)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P19975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19975G 5 g
P199725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19975G Supplier TCI America Supplier No. P19975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30131-16-9
Molecular Formula C17H18O3
Molecular Weight (g/mol) 270.33
MDL Number MFCD07787608
InChI Key XWCWFMQMZZPALR-UHFFFAOYSA-N
PubChem CID 11043825
IUPAC Name 4-(4-phenylbutoxy)benzoic acid
SMILES OC(=O)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1

Specifications

Melting Point 131°C
Color White
Quantity 5 g
Formula Weight 270.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Phenylbutoxy)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.