CAS RN 301359-91-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ eIF-2α Inhibitor II, Sal003
The eIF-2α Inhibitor II, Sal003, also referenced under CAS 301359-91-1, controls the biological activity of eIF-2α. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.