accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3017-60-5

IUPAC Name: λ²-cobalt(2+) bis(cyanosulfanide)
Synonyms: cobalt(II) thiocyanate
Molecular Weight (g/mol): 175.09
Molecular Formula: C2CoN2S2
InChi Key: INDBQWVYFLTCFF-UHFFFAOYSA-L
SMILES: [Co++].[S-]C#N.[S-]C#N
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (5)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)