Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Thiocyanate, MP Biomedicals

Catalog No. ICN214178 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214178 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214178 Supplier MP Biomedicals, Inc Supplier No. 0521417810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS grade chemical for research and further manufacturing use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3017-60-5
Molecular Formula C2H2CoN2S2
Molecular Weight (g/mol) 177.105
InChI Key IQHZUTZWNWRODJ-UHFFFAOYSA-N
PubChem CID 131877393
IUPAC Name cobalt;thiocyanic acid
SMILES C(#N)S.C(#N)S.[Co]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 175.1
Chemical Name or Material Cobaltous Thiocyanate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.