CAS RN 3017-95-6
IUPAC Name:
2-bromo-1-chloropropane
Synonyms:
2-bromo-1-chloropropane
Molecular Weight (g/mol):
157.44
Molecular Formula:
C3H6BrCl
InChi Key:
PUJJZGFFAQHYEX-UHFFFAOYNA-N
SMILES:
CC(Br)CCl
1
–
1
of
1
results
1
2-Bromo-1-Chlropropane, SPEX CertiPrep™
CAS: 3017-95-6 Molecular Formula: C3H6BrCl Molecular Weight (g/mol): 157.435 InChI Key: PUJJZGFFAQHYEX-UHFFFAOYSA-N PubChem CID: 18175 IUPAC Name: 2-bromo-1-chloropropane SMILES: CC(CCl)Br